Assay Details
Target Gene Details
Entrez Gene ID: | 55296 |
Gene Name: | TBC1 domain family member 19 |
Gene Aliases: |
- |
Location: |
Chr.4:26583924-26858919 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D19 | NM_001292054.1 | 5 | 612 | NP_001278983.1 |
| NM_018317.3 | 8 | 807 | NP_060787.2 | |
| XM_011513852.2 | 8 | 647 | XP_011512154.1 | |
| XM_017008363.1 | 8 | 647 | XP_016863852.1 | |
| XM_017008364.1 | 8 | 647 | XP_016863853.1 | |
| AB449905.1 | 5 | 342 | ||
| AK001944.1 | 8 | 647 | BAA91992.1 | |
| AK225395.1 | 8 | 647 | ||
| AK289631.1 | 8 | 647 | ||
| AK310795.1 | 8 | 654 | ||
| BC031642.1 | 8 | 603 | AAH31642.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522423 | Chr.4:26623563 - 26704720 on Build GRCh38 | Gain |
|
| nsv524115 | Chr.4:26629556 - 26704720 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map