Assay Details
Target Gene Details
Entrez Gene ID: | 2617 |
Gene Name: | glycyl-tRNA synthetase |
Gene Aliases: |
CMT2D, DSMAV, GlyRS, HMN5, SMAD1 |
Location: |
Chr.7:30594735-30634033 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GARS | NM_001316772.1 | NP_001303701.1 | ||
| NM_002047.3 | NP_002038.2 | |||
| XM_006715686.2 | XP_006715749.1 | |||
| AK074524.1 | ||||
| AK295490.1 | ||||
| AK310351.1 | ||||
| BC007722.2 | AAH07722.1 | |||
| BC007755.2 | AAH07755.1 | |||
| D30658.1 | BAA06338.1 | |||
| U09510.1 | AAA86443.1 | |||
| U09587.1 | AAA57001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024458 | Chr.7:30562261 - 30610043 on Build GRCh38 | Loss |
|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map