Assay Details
Target Gene Details
Entrez Gene ID: | 138802 |
Gene Name: | olfactory receptor family 13 subfamily C member 8 |
Gene Aliases: |
OR37H, OR9-10 |
Location: |
Chr.9:104569168-104570130 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR13C8 | NM_001004483.1 | 1 | 114 | NP_001004483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831677 | Chr.9:104519297 - 104688058 on Build GRCh38 | Gain |
|
| esv3891706 | Chr.9:104480016 - 105304573 on Build GRCh38 | Gain |
|
| esv3891705 | Chr.9:104391828 - 106660921 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map