Assay Details
Target Gene Details
Entrez Gene ID: | 51388 |
Gene Name: | NIP7, nucleolar pre-rRNA processing protein |
Gene Aliases: |
CGI-37, HSPC031, KD93 |
Location: |
Chr.16:69339512-69343111 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIP7 | NM_001199434.1 | 1 | 285 | NP_001186363.1 |
| NM_016101.4 | 1 | 285 | NP_057185.1 | |
| AB062398.1 | BAB93485.1 | |||
| AB062479.1 | BAB93504.1 | |||
| AB112439.2 | 1 | 125 | BAD05056.1 | |
| AF085360.1 | AAD40195.1 | |||
| AF132971.1 | AAD27746.1 | |||
| AF161528.1 | AAF29143.1 | |||
| AK001158.1 | ||||
| AK315355.1 | ||||
| BC015941.1 | AAH15941.1 | |||
| BM754629.1 | 1 | 154 | ||
| DB453898.1 | 1 | 369 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827715 | Chr.16:69311943 - 69340458 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map