Assay Details
Target Gene Details
Entrez Gene ID: | 8643 |
Gene Name: | patched 2 |
Gene Aliases: |
PTC2 |
Location: |
Chr.1:44819844-44842944 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTCH2 | NM_001166292.1 | 12 | 1547 | NP_001159764.1 |
| NM_003738.4 | 12 | 1547 | NP_003729.3 | |
| AF087651.1 | 12 | 1535 | AAD25953.1 | |
| AF091501.1 | 12 | 1535 | AAC79847.1 | |
| AF119569.1 | 12 | 1547 | AAD17260.1 | |
| AK307168.1 | 1 | 264 | ||
| AY358555.1 | 12 | 1535 | AAQ88919.1 | |
| AY359016.1 | 12 | 1539 | AAQ89375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546146 | Chr.1:44792469 - 44853080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map