Assay Details
Target Gene Details
Entrez Gene ID: | 23416 |
Gene Name: | potassium voltage-gated channel subfamily H member 3 |
Gene Aliases: |
BEC1, ELK2, Kv12.2 |
Location: |
Chr.12:49538541-49558312 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH3 | NM_001314030.1 | NP_001300959.1 | ||
| NM_012284.2 | NP_036416.1 | |||
| XM_011538085.2 | XP_011536387.1 | |||
| XM_011538086.2 | XP_011536388.1 | |||
| XM_017019096.1 | XP_016874585.1 | |||
| XM_017019097.1 | XP_016874586.1 | |||
| AB022696.1 | BAA83590.1 | |||
| AB033108.1 | ||||
| BC150602.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map