Assay Details
Target Gene Details
Entrez Gene ID: | 9166 |
Gene Name: | estrogen receptor binding site associated, antigen, 9 |
Gene Aliases: |
EB9, PDAF |
Location: |
Chr.8:109539700-109565996 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBAG9 | NM_004215.4 | NP_004206.1 | ||
| NM_198120.2 | NP_936056.1 | |||
| AK290651.1 | ||||
| BC017729.1 | AAH17729.1 | |||
| BG427211.1 | ||||
| BG707115.1 | ||||
| BI194881.1 | ||||
| CD108096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1075574 | Chr.8:109539370 - 109540171 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map