Assay Details
Target Gene Details
Entrez Gene ID: | 285598 |
Gene Name: | ADP ribosylation factor like GTPase 10 |
Gene Aliases: |
ARL10A |
Location: |
Chr.5:176365468-176415085 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL10 | NM_173664.5 | 4 | 1022 | NP_775935.1 |
| XM_011534529.2 | XP_011532831.1 | |||
| XM_011534530.2 | XP_011532832.1 | |||
| XM_011534531.2 | XP_011532833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023853 | Chr.5:176043309 - 176463537 on Build GRCh38 | Gain |
|
| nsv600341 | Chr.5:176132836 - 176384132 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map