Assay Details
Target Gene Details
Entrez Gene ID: | 526 |
Gene Name: | ATPase H+ transporting V1 subunit B2 |
Gene Aliases: |
ATP6B1B2, ATP6B2, DOOD, HO57, VATB, VPP3, Vma2, ZLS2 |
Location: |
Chr.8:20197193-20226852 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1B2 | NM_001693.3 | NP_001684.2 | ||
| AB208806.1 | BAD92043.1 | |||
| AK294163.1 | ||||
| AK298819.1 | ||||
| AK312372.1 | ||||
| BC003100.1 | AAH03100.1 | |||
| BC007309.1 | AAH07309.1 | |||
| BC030640.2 | AAH30640.1 | |||
| L35249.1 | AAA58661.1 | |||
| M60346.1 | AAA35610.1 | |||
| X62949.1 | CAA44721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610759 | Chr.8:19704486 - 20211331 on Build GRCh38 | Gain |
|
| nsv522185 | Chr.8:19097292 - 20497802 on Build GRCh38 | Gain |
|
| nsv1033511 | Chr.8:19893788 - 20767580 on Build GRCh38 | Gain |
|
| nsv6107 | Chr.8:20198643 - 20243283 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map