Assay Details
Target Gene Details
Entrez Gene ID: | 5891 |
Gene Name: | MOK protein kinase |
Gene Aliases: |
RAGE, RAGE-1, RAGE1, STK30 |
Location: |
Chr.14:102224450-102305200 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22037 | Chr.14:102223226 - 102248955 on Build GRCh38 | Gain |
|
| nsv1429 | Chr.14:102201072 - 102246584 on Build GRCh38 | Deletion |
|
| esv3635547 | Chr.14:102223491 - 102317911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map