Assay Details
Target Gene Details
Entrez Gene ID: | 79170 |
Gene Name: | proline rich 15 like |
Gene Aliases: |
ATAD4 |
Location: |
Chr.17:47951967-47957905 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR15L | NM_024320.3 | 1 | 156 | NP_077296.1 |
| XM_005257663.4 | 1 | 183 | XP_005257720.1 | |
| XM_005257666.4 | 1 | 184 | XP_005257723.1 | |
| AK309589.1 | 1 | 127 | ||
| BC002865.2 | AAH02865.1 | |||
| BM829856.1 | 1 | 156 |

Genomic Map