Assay Details
Target Gene Details
Entrez Gene ID: | 115727 |
Gene Name: | RAS guanyl releasing protein 4 |
Gene Aliases: |
- |
Location: |
Chr.19:38409055-38426305 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASGRP4 | NM_001146202.1 | 17 | 2303 | NP_001139674.1 |
| NM_001146203.1 | 16 | 2069 | NP_001139675.1 | |
| NM_001146204.1 | 17 | 2138 | NP_001139676.1 | |
| NM_001146205.1 | 17 | 2243 | NP_001139677.1 | |
| NM_001146206.1 | 14 | 1778 | NP_001139678.1 | |
| NM_001146207.1 | 15 | 2054 | NP_001139679.1 | |
| NM_170604.2 | 17 | 2345 | NP_733749.1 | |
| AF448437.1 | 17 | 2202 | AAL87858.1 | |
| AK097292.1 | 9 | 1416 | ||
| AY048119.1 | AAK85701.1 | |||
| AY048120.1 | AAK85702.1 | |||
| AY048121.1 | AAK85703.1 | |||
| BC146669.1 | 17 | 2185 | ||
| BC150202.1 | 17 | 2185 | ||
| FJ768677.1 | 16 | 2069 | ||
| FJ768678.1 | 17 | 2138 | ||
| FJ768679.1 | 17 | 2243 | ||
| FJ768680.1 | 14 | 1778 | ||
| FJ768681.1 | 16 | 2440 | ||
| FJ768682.1 | 15 | 2054 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|
| nsv833823 | Chr.19:38364910 - 38545160 on Build GRCh38 | Loss |
|
| dgv3525n100 | Chr.19:38405604 - 38459089 on Build GRCh38 | Gain |
|
| nsv470140 | Chr.19:38349499 - 38628854 on Build GRCh38 | Gain |
|
| nsv1064275 | Chr.19:38369225 - 38422611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map