Assay Details
Target Gene Details
Entrez Gene ID: | 10197 |
Gene Name: | proteasome activator subunit 3 |
Gene Aliases: |
HEL-S-283, Ki, PA28-gamma, PA28G, PA28gamma, REG-GAMMA |
Location: |
Chr.17:42833142-42843760 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSME3 | NM_001267045.1 | 13 | 1737 | NP_001253974.1 |
| NM_005789.3 | 11 | 1410 | NP_005780.2 | |
| NM_176863.2 | 11 | 1449 | NP_789839.1 | |
| NR_049772.1 | 10 | 1297 | ||
| XM_011524177.2 | 11 | 1419 | XP_011522479.1 | |
| AK057221.1 | 13 | 1576 | ||
| AK074999.1 | 11 | 1107 | ||
| AK303211.1 | ||||
| BC001423.2 | 11 | 1137 | AAH01423.1 | |
| BC002684.1 | AAH02684.1 | |||
| BC008020.1 | 11 | 1068 | AAH08020.1 | |
| BX640801.1 | 13 | 1466 | CAE45884.1 | |
| EU794653.1 | 11 | 1137 | ||
| U11292.1 | 11 | 1121 | AAB60335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map