Assay Details
Target Gene Details
Entrez Gene ID: | 80777 |
Gene Name: | cytochrome b5 type B |
Gene Aliases: |
CYB5-M, CYPB5M, OMB5 |
Location: |
Chr.16:69424595-69466264 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5B | NM_030579.2 | NP_085056.2 | ||
| AB009282.1 | BAA23735.1 | |||
| AK291576.1 | ||||
| BC004373.1 | AAH04373.1 | |||
| BC014431.1 | AAH14431.2 | |||
| BX647922.1 | 3 | 411 | CAI46070.1 |
More Information
Set Membership: |
|

Genomic Map