Assay Details
Target Gene Details
Entrez Gene ID: | 5879 |
Gene Name: | ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) |
Gene Aliases: |
MIG5, Rac-1, TC-25, p21-Rac1 |
Location: |
Chr.7:6374495-6403967 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAC1 | NM_006908.4 | NP_008839.2 | ||
| NM_018890.3 | NP_061485.1 | |||
| AK054993.1 | ||||
| BC050687.1 | AAH50687.1 | |||
| D25274.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020236 | Chr.7:6209299 - 6650746 on Build GRCh38 | Gain |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv605999 | Chr.7:6357088 - 6433250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map