Assay Details
Target Gene Details
Entrez Gene ID: | 55850 |
Gene Name: | unconventional SNARE in the ER 1 |
Gene Aliases: |
D12, MDS032, P31, SLT1 |
Location: |
Chr.19:17214528-17219831 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USE1 | NM_018467.3 | 7 | 572 | NP_060937.1 |
| XM_017026975.1 | 8 | 1282 | XP_016882464.1 | |
| XM_017026976.1 | 6 | 522 | XP_016882465.1 | |
| AB097050.1 | 7 | 543 | BAC77403.1 | |
| AF220052.1 | 7 | 543 | AAF67645.1 | |
| BC006005.1 | AAH06005.1 | |||
| BC008455.1 | 7 | 520 | AAH08455.1 | |
| BF448659.1 | 2 | 287 | ||
| BI758045.1 | 7 | 559 | ||
| T86451.1 | 2 | 286 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458373 | Chr.19:17183487 - 17226535 on Build GRCh38 | Loss |
|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map