Assay Details
Target Gene Details
Entrez Gene ID: | 84270 |
Gene Name: | caspase recruitment domain family member 19 |
Gene Aliases: |
BinCARD, C9orf89 |
Location: |
Chr.9:93096123-93113295 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD19 | NM_001318010.1 | 4 | 533 | NP_001304939.1 |
| NM_001318011.1 | 5 | 676 | NP_001304940.1 | |
| NM_032310.4 | 6 | 736 | NP_115686.3 | |
| NR_134464.1 | 4 | 2183 | ||
| NR_134465.1 | 4 | 2172 | ||
| NR_134466.1 | 6 | 747 | ||
| NR_134467.1 | 7 | 838 | ||
| NR_134468.1 | 5 | 687 | ||
| AA804861.1 | 1 | 216 | ||
| AK057716.1 | 4 | 2077 | BAB71550.1 | |
| AK091611.1 | 4 | 2088 | ||
| AK097977.1 | 2 | 2474 | ||
| AK128259.1 | 2 | 3209 | ||
| AW207349.1 | 1 | 232 | ||
| BC004500.2 | 7 | 605 | AAH04500.1 | |
| BM930683.1 | 5 | 521 | ||
| BX354102.2 | 7 | 718 | ||
| CD676329.1 | 5 | 543 | ||
| DB550397.1 | 4 | 232 | ||
| DN998463.1 | 4 | 426 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526296 | Chr.9:92945910 - 93268110 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map