Assay Details
Target Gene Details
Entrez Gene ID: | 4713 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B7 |
Gene Aliases: |
B18, CI-B18 |
Location: |
Chr.19:14566078-14572077 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB7 | NM_004146.5 | 2 | 271 | NP_004137.2 |
| XM_011528039.2 | 2 | 863 | XP_011526341.1 | |
| AF112200.1 | 2 | 257 | AAF17188.1 | |
| AF217091.1 | 2 | 216 | AAF91223.1 | |
| BC002595.2 | 2 | 256 | AAH02595.1 | |
| CB124371.1 | 2 | 271 | ||
| CR450354.1 | 2 | 194 | CAG29350.1 | |
| M33374.1 | 2 | 229 | AAA35675.1 |
More Information
Set Membership: |
|

Genomic Map