Assay Details
Target Gene Details
Entrez Gene ID: | 23376 |
Gene Name: | UFM1 specific ligase 1 |
Gene Aliases: |
KIAA0776, Maxer, NLBP, RCAD |
Location: |
Chr.6:96521826-96555276 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UFL1 | NM_015323.4 | NP_056138.1 | ||
| AB018319.1 | ||||
| AK295934.1 | ||||
| AK307651.1 | ||||
| AK316453.1 | ||||
| BC015377.1 | ||||
| BC036379.2 | AAH36379.1 | |||
| BP337693.1 |
More Information
Set Membership: |
|

Genomic Map