Assay Details
Target Gene Details
Entrez Gene ID: | 127343 |
Gene Name: | diencephalon/mesencephalon homeobox 1 |
Gene Aliases: |
MBX, OTX3, PAXB |
Location: |
Chr.1:46489837-46514226 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMBX1 | NM_147192.2 | NP_671725.1 | ||
| NM_172225.1 | NP_757379.1 | |||
| XM_011540668.2 | XP_011538970.1 | |||
| XM_017000289.1 | XP_016855778.1 | |||
| AB037699.1 | BAC00920.1 | |||
| AF398527.1 | AAM90589.1 | |||
| AF398528.1 | AAM90590.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009193 | Chr.1:46474327 - 46619995 on Build GRCh38 | Gain |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map