Assay Details
Target Gene Details
Entrez Gene ID: | 7976 |
Gene Name: | frizzled class receptor 3 |
Gene Aliases: |
Fz-3 |
Location: |
Chr.8:28494205-28574268 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FZD3 | NM_017412.3 | 3 | 235 | NP_059108.1 |
| NM_145866.1 | 2 | 189 | NP_665873.1 | |
| XM_017013842.1 | 3 | 225 | XP_016869331.1 | |
| XM_017013843.1 | 3 | 225 | XP_016869332.1 | |
| XM_017013844.1 | 3 | 225 | XP_016869333.1 | |
| AB039723.1 | 3 | 186 | BAA94968.1 | |
| AJ272427.1 | CAB89114.1 | |||
| AK291480.1 | 2 | 184 | ||
| DA250029.1 | 3 | 232 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7157n100 | Chr.8:28499087 - 28584581 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map