Assay Details
Target Gene Details
Entrez Gene ID: | 56954 |
Gene Name: | nitrilase family member 2 |
Gene Aliases: |
HEL-S-8a |
Location: |
Chr.3:100334718-100355634 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIT2 | NM_020202.4 | NP_064587.1 | ||
| AF260334.1 | AAG44665.1 | |||
| AF284574.1 | AAF87103.1 | |||
| AK225979.1 | ||||
| AK295835.1 | 6 | 526 | ||
| AK313704.1 | ||||
| BC020620.1 | AAH20620.1 | |||
| BC107890.1 | AAI07891.1 | |||
| FJ224326.1 |
More Information
Set Membership: |
|

Genomic Map