Assay Details
Target Gene Details
Entrez Gene ID: | 161514 |
Gene Name: | TBC1 domain family member 21 |
Gene Aliases: |
MgcRabGAP |
Location: |
Chr.15:73873556-73909783 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D21 | NM_001286434.1 | NP_001273363.1 | ||
| NM_153356.2 | NP_699187.1 | |||
| XM_006720409.3 | XP_006720472.1 | |||
| XM_006720410.3 | XP_006720473.1 | |||
| XM_006720411.3 | XP_006720474.1 | |||
| XM_011521281.2 | XP_011519583.1 | |||
| XM_011521282.2 | XP_011519584.1 | |||
| XM_011521283.2 | XP_011519585.1 | |||
| XM_011521284.2 | XP_011519586.1 | |||
| AB449907.1 | ||||
| BC033516.1 | AAH33516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map