Assay Details
Target Gene Details
Entrez Gene ID: | 153 |
Gene Name: | adrenoceptor beta 1 |
Gene Aliases: |
ADRB1R, B1AR, BETA1AR, RHR |
Location: |
Chr.10:114044047-114046908 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRB1 | NM_000684.2 | 1 | 1876 | NP_000675.1 |
| AF272890.1 | 1 | 153 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552203 | Chr.10:114041494 - 114047023 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map