Assay Details
Target Gene Details
Entrez Gene ID: | 158297 |
Gene Name: | stabilizer of axonemal microtubules 1 |
Gene Aliases: |
C9orf138, FAM154A |
Location: |
Chr.9:18927660-19049354 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAXO1 | NM_001287049.1 | 4 | 1081 | NP_001273978.1 |
| NM_001287050.1 | 3 | 1120 | NP_001273979.1 | |
| NM_153707.3 | 4 | 1323 | NP_714918.2 | |
| XM_011517744.1 | 5 | 1403 | XP_011516046.1 | |
| XM_011517745.1 | 6 | 1338 | XP_011516047.1 | |
| XM_011517746.2 | 6 | 1259 | XP_011516048.1 | |
| XM_011517747.2 | 3 | 919 | XP_011516049.1 | |
| XM_017014314.1 | 6 | 1649 | XP_016869803.1 | |
| XM_017014315.1 | 4 | 1036 | XP_016869804.1 | |
| AK057474.1 | 3 | 1115 | ||
| AK292245.1 | 4 | 1274 | ||
| BC033489.1 | 4 | 1251 | AAH33489.1 | |
| BM563230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525659 | Chr.9:18288214 - 19009043 on Build GRCh38 | Gain |
|
| nsv519384 | Chr.9:18924138 - 18931063 on Build GRCh38 | Loss |
|
| esv3619882 | Chr.9:18831160 - 18952565 on Build GRCh38 | Gain |
|
| nsv471289 | Chr.9:18920359 - 18960395 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map