Assay Details
Target Gene Details
Entrez Gene ID: | 375323 |
Gene Name: | lipoma HMGIC fusion partner-like 4 |
Gene Aliases: |
- |
Location: |
Chr.3:9498361-9553802 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL4 | NM_198560.2 | 4 | 4500 | NP_940962.1 |
| XM_017006359.1 | 3 | 5038 | XP_016861848.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834614 | Chr.3:9495688 - 9639466 on Build GRCh38 | Gain |
|

Genomic Map