Assay Details
Target Gene Details
Entrez Gene ID: | 113177 |
Gene Name: | IZUMO family member 4 |
Gene Aliases: |
C19orf36, IMAGE:4215339 |
Location: |
Chr.19:2096848-2099590 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IZUMO4 | NM_001031735.2 | 9 | 719 | NP_001026905.2 |
| NM_001039846.1 | 10 | 773 | NP_001034935.1 | |
| XM_005259480.3 | 8 | 722 | XP_005259537.2 | |
| AW182303.1 | 1 | 248 | ||
| AY358444.1 | 7 | 1892 | AAQ88809.1 | |
| AY753639.1 | ||||
| BC010443.1 | 6 | 1603 | AAH10443.1 | |
| BC014609.2 | 9 | 673 | AAH14609.1 | |
| BI827974.1 | 10 | 628 | ||
| BX378854.2 | 10 | 773 | ||
| DQ196309.1 | 9 | 706 | ||
| DQ196310.1 | 10 | 760 | ||
| DQ424900.1 | ABD83665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss |
|
| nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss |
|
| esv3583205 | Chr.19:2096468 - 2114273 on Build GRCh38 | Loss |
|

Genomic Map