Assay Details
Target Gene Details
Entrez Gene ID: | 7225 |
Gene Name: | transient receptor potential cation channel subfamily C member 6 |
Gene Aliases: |
FSGS2, TRP6 |
Location: |
Chr.11:101451470-101584018 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC6 | NM_004621.5 | 13 | 3521 | NP_004612.2 |
| XM_011542968.2 | 13 | 3128 | XP_011541270.1 | |
| XM_017018221.1 | 11 | 3263 | XP_016873710.1 | |
| XM_017018222.1 | 13 | 3023 | XP_016873711.1 | |
| AF080394.2 | AAC63289.2 | |||
| AJ006276.1 | 13 | 3523 | CAA06943.1 | |
| AK001960.1 | 3 | 625 | ||
| AK027769.1 | 12 | 2166 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468851 | Chr.11:101397716 - 101460076 on Build GRCh38 | Gain |
|
| nsv556131 | Chr.11:101115046 - 101609286 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map