Assay Details
Target Gene Details
Entrez Gene ID: | 84056 |
Gene Name: | katanin catalytic subunit A1 like 1 |
Gene Aliases: |
- |
Location: |
Chr.13:30202626-30307500 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNAL1 | NM_001014380.2 | 11 | 1489 | NP_001014402.1 |
| NM_032116.4 | 11 | 1531 | NP_115492.1 | |
| XM_005266574.2 | 11 | 1673 | XP_005266631.1 | |
| XM_017020791.1 | 10 | 1375 | XP_016876280.1 | |
| XM_017020792.1 | 10 | 1396 | XP_016876281.1 | |
| XM_017020793.1 | 8 | 1705 | XP_016876282.1 | |
| AK097423.1 | 11 | 1461 | ||
| AK291439.1 | 11 | 1464 | ||
| AK315724.1 | 11 | 1468 | ||
| BC000612.2 | 11 | 1491 | AAH00612.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631670 | Chr.13:30207544 - 30210485 on Build GRCh38 | Loss |
|
| esv2657382 | Chr.13:30207530 - 30210476 on Build GRCh38 | Deletion |
|
| esv3580589 | Chr.13:30207962 - 30215532 on Build GRCh38 | Loss |
|
| esv3631667 | Chr.13:30146805 - 30231044 on Build GRCh38 | Gain |
|
| nsv832572 | Chr.13:30075868 - 30261119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map