Assay Details
Target Gene Details
Entrez Gene ID: | 23170 |
Gene Name: | tubulin tyrosine ligase like 12 |
Gene Aliases: |
dJ526I14.2 |
Location: |
Chr.22:43166622-43187131 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL12 | NM_015140.3 | NP_055955.1 | ||
| AL590888.1 | CAC37415.1 | |||
| BC001070.2 | AAH01070.1 | |||
| CT841516.1 | CAJ86446.1 | |||
| D63487.1 | BAA09774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762124 | Chr.22:43031130 - 43188670 on Build GRCh38 | Gain |
|
| nsv520927 | Chr.22:43154143 - 43307985 on Build GRCh38 | Loss |
|
| nsv459890 | Chr.22:43152149 - 43183316 on Build GRCh38 | Gain |
|
| esv33816 | Chr.22:43113758 - 43326122 on Build GRCh38 | Gain |
|
| nsv834214 | Chr.22:43037931 - 43217928 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map