Assay Details
Target Gene Details
Entrez Gene ID: | 1482 |
Gene Name: | NK2 homeobox 5 |
Gene Aliases: |
CHNG5, CSX, CSX1, HLHS2, NKX2.5, NKX2E, NKX4-1, VSD3 |
Location: |
Chr.5:173232104-173235312 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-5 | NM_001166175.1 | 2 | 1533 | NP_001159647.1 |
| NM_001166176.1 | 2 | 1496 | NP_001159648.1 | |
| NM_004387.3 | 2 | 1241 | NP_004378.1 | |
| AB021133.1 | BAA35181.1 | |||
| AI609745.1 | 1 | 429 | ||
| AK290615.1 | 2 | 1133 | ||
| BC025711.1 | 2 | 1119 | AAH25711.1 | |
| KU178001.1 | ||||
| KU178002.1 | ||||
| U34962.1 | 2 | 1188 | AAC50470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950444 | Chr.5:173228498 - 173240197 on Build GRCh38 | Deletion |
|
| esv3575898 | Chr.5:173222243 - 173235416 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map