Assay Details
Target Gene Details
Entrez Gene ID: | 644096 |
Gene Name: | succinate dehydrogenase complex assembly factor 1 |
Gene Aliases: |
LYRM8 |
Location: |
Chr.19:35995188-35996318 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHAF1 | NM_001042631.2 | 1 | 461 | NP_001036096.1 |
| BC137516.1 | 1 | 404 | ||
| BC137517.1 | 1 | 404 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv51n68 | Chr.19:35841011 - 36016692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map