Assay Details
Target Gene Details
Entrez Gene ID: | 60559 |
Gene Name: | signal peptidase complex subunit 3 |
Gene Aliases: |
PRO3567, SPC22/23, SPC3, YLR066W |
Location: |
Chr.4:176319939-176332245 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPCS3 | NM_021928.3 | 1 | 218 | NP_068747.1 |
| AK026302.1 | 1 | 193 | BAB15437.1 | |
| AL136660.1 | 1 | 188 | CAB66595.1 | |
| AY359044.1 | 1 | 176 | AAQ89403.1 | |
| BC047290.1 | 1 | 161 | AAH47290.1 | |
| DB285025.1 | 1 | 219 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894072 | Chr.4:176311557 - 176329340 on Build GRCh38 | Loss |
|
| esv34137 | Chr.4:175851071 - 176341258 on Build GRCh38 | Loss |
|
| esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion |
|
| esv3603274 | Chr.4:176144758 - 176386481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map