Assay Details
Target Gene Details
Entrez Gene ID: | 56158 |
Gene Name: | testis expressed 12 |
Gene Aliases: |
- |
Location: |
Chr.11:112167372-112172556 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX12 | NM_031275.4 | NP_112565.1 | ||
| AF285600.1 | AAK31979.1 | |||
| BC029506.1 | AAH29506.1 | |||
| BI561514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
| nsv519192 | Chr.11:112160901 - 112193416 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map