Assay Details
Target Gene Details
Entrez Gene ID: | 7704 |
Gene Name: | zinc finger and BTB domain containing 16 |
Gene Aliases: |
PLZF, ZNF145 |
Location: |
Chr.11:114059576-114256765 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB16 | NM_006006.4 | NP_005997.2 | ||
| XM_005271658.4 | 1 | 154 | XP_005271715.1 | |
| AB208916.1 | BAD92153.1 | |||
| BC029812.1 | AAH29812.1 | |||
| BX648973.1 | 1 | 139 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952735 | Chr.11:114058779 - 114061778 on Build GRCh38 | Deletion |
|
| nsv826091 | Chr.11:114058394 - 114063044 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map