Assay Details
Target Gene Details
Entrez Gene ID: | 8785 |
Gene Name: | matrilin 4 |
Gene Aliases: |
- |
Location: |
Chr.20:45293446-45308566 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MATN4 | NM_003833.4 | NP_003824.2 | ||
| NM_030590.3 | NP_085080.1 | |||
| NM_030592.3 | NP_085095.1 | |||
| XM_005260597.1 | XP_005260654.1 | |||
| XM_017028113.1 | XP_016883602.1 | |||
| XM_017028114.1 | XP_016883603.1 | |||
| XM_017028115.1 | XP_016883604.1 | |||
| AJ007581.1 | CAA07569.1 | |||
| AK074495.1 | ||||
| AK074593.1 | ||||
| AK074595.1 | BAC11081.1 | |||
| AK074597.1 | BAC11083.1 | |||
| BC131763.1 | ||||
| BC141811.1 | ||||
| BC151219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056815 | Chr.20:45257396 - 45314077 on Build GRCh38 | Loss |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
| nsv1057759 | Chr.20:45283246 - 45307625 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map