Assay Details
Target Gene Details
Entrez Gene ID: | 29911 |
Gene Name: | hook microtubule-tethering protein 2 |
Gene Aliases: |
HK2 |
Location: |
Chr.19:12763002-12778484 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOOK2 | NM_001100176.1 | NP_001093646.1 | ||
| NM_013312.2 | NP_037444.2 | |||
| XM_005259873.2 | XP_005259930.1 | |||
| XM_005259874.2 | XP_005259931.1 | |||
| XM_005259876.4 | XP_005259933.1 | |||
| XM_005259877.4 | XP_005259934.1 | |||
| XM_011527943.2 | XP_011526245.1 | |||
| XM_011527944.2 | XP_011526246.1 | |||
| XM_011527945.2 | XP_011526247.1 | |||
| XM_011527946.2 | XP_011526248.1 | |||
| XM_011527947.2 | XP_011526249.1 | |||
| XM_017026720.1 | XP_016882209.1 | |||
| XM_017026721.1 | XP_016882210.1 | |||
| AF044924.1 | AAC09299.1 | |||
| BC012443.1 | AAH12443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv1160586 | Chr.19:12597480 - 12823909 on Build GRCh38 | Duplication |
|
| dgv138n111 | Chr.19:12627853 - 12810372 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map