Assay Details
Target Gene Details
Entrez Gene ID: | 26257 |
Gene Name: | NK2 homeobox 8 |
Gene Aliases: |
NKX2.8, NKX2H, Nkx2-9 |
Location: |
Chr.14:36580011-36582614 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-8 | NM_014360.3 | 2 | 1501 | NP_055175.2 |
| BC041090.1 | 2 | 1468 | AAH41090.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32575 | Chr.14:36578424 - 36583453 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74949744] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map