Assay Details
Target Gene Details
Entrez Gene ID: | 84101 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
- |
Location: |
Chr.12:95516560-95551529 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580379 | Chr.12:95518596 - 95528911 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map