Assay Details
Target Gene Details
Entrez Gene ID: | 7185 |
Gene Name: | TNF receptor associated factor 1 |
Gene Aliases: |
EBI6, MGC:10353 |
Location: |
Chr.9:120902393-120929173 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF1 | NM_001190945.1 | 9 | 2006 | NP_001177874.1 |
| NM_001190947.1 | 6 | 1418 | NP_001177876.1 | |
| NM_005658.4 | 8 | 2153 | NP_005649.1 | |
| AK090468.1 | 8 | 2154 | BAC03449.1 | |
| AL832989.1 | 8 | 4026 | CAH56343.1 | |
| BC024145.2 | 7 | 1659 | AAH24145.1 | |
| U19261.1 | 7 | 1656 | AAA62309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759713 | Chr.9:120885919 - 121080717 on Build GRCh38 | Loss |
|
| nsv6700 | Chr.9:120896969 - 120930014 on Build GRCh38 | Insertion |
|
| nsv1051370 | Chr.9:120810573 - 120906638 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map