Assay Details
Target Gene Details
Entrez Gene ID: | 55298 |
Gene Name: | ring finger protein 121 |
Gene Aliases: |
- |
Location: |
Chr.11:71928701-71997597 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF121 | NM_001300926.1 | 7 | 2198 | NP_001287855.1 |
| NM_018320.4 | 9 | 2360 | NP_060790.2 | |
| NR_024147.1 | 8 | 2322 | ||
| NR_024148.1 | 9 | 2460 | ||
| XM_006718629.1 | 8 | 2273 | XP_006718692.1 | |
| XM_006718630.1 | 10 | 2173 | XP_006718693.1 | |
| XM_006718632.1 | 7 | 2118 | XP_006718695.1 | |
| XM_011545150.1 | 10 | 2205 | XP_011543452.1 | |
| AI291325.1 | 1 | 244 | ||
| AK001961.1 | 4 | 1524 | BAA92002.1 | |
| AK023139.1 | 9 | 2026 | BAB14423.1 | |
| AK094508.1 | 9 | 2126 | ||
| AL833638.1 | 7 | 1873 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map