Assay Details
Target Gene Details
Entrez Gene ID: | 51562 |
Gene Name: | MAP3K12 binding inhibitory protein 1 |
Gene Aliases: |
- |
Location: |
Chr.14:36298558-36320676 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBIP | NM_001144891.1 | 9 | 1057 | NP_001138363.1 |
| NM_001308110.1 | 8 | 962 | NP_001295039.1 | |
| NM_016586.2 | 9 | 1060 | NP_057670.2 | |
| XM_005267754.3 | 8 | 989 | XP_005267811.1 | |
| XM_005267756.4 | 8 | 954 | XP_005267813.1 | |
| XM_017021367.1 | 7 | 888 | XP_016876856.1 | |
| AB038523.1 | 9 | 1029 | BAA94083.1 | |
| AF208857.1 | 10 | 1228 | AAF64271.1 | |
| AK293473.1 | 9 | 941 | ||
| AK315290.1 | 9 | 1061 | ||
| BC005197.1 | 9 | 1019 | AAH05197.1 | |
| BC016821.1 | 9 | 1012 | AAH16821.1 | |
| BX248287.1 | 8 | 882 | CAD62615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv83e55 | Chr.14:36267143 - 36313743 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map