Assay Details
Target Gene Details
Entrez Gene ID: | 2947 |
Gene Name: | glutathione S-transferase mu 3 |
Gene Aliases: |
GST5, GSTB, GSTM3-3, GTM3 |
Location: |
Chr.1:109733932-109741038 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSTM3 | NM_000849.4 | NP_000840.2 | ||
| NR_024537.1 | ||||
| AB209816.1 | BAD93053.1 | |||
| AK304300.1 | ||||
| AK313444.1 | ||||
| BC000088.2 | AAH00088.1 | |||
| BC008790.2 | AAH08790.1 | |||
| BT019945.1 | AAV38748.1 | |||
| CR542108.1 | CAG46905.1 | |||
| EF363096.1 | ||||
| J05459.1 | AAA60964.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003914 | Chr.1:109684109 - 109741677 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map