Assay Details
Target Gene Details
Entrez Gene ID: | 167555 |
Gene Name: | family with sequence similarity 151 member B |
Gene Aliases: |
UNQ9217 |
Location: |
Chr.5:80487981-80542563 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM151B | NM_205548.2 | 4 | 550 | NP_991111.2 |
| XM_006714564.3 | 4 | 468 | XP_006714627.2 | |
| XM_011543234.2 | 4 | 929 | XP_011541536.1 | |
| XM_011543235.2 | 3 | 304 | XP_011541537.1 | |
| XM_011543236.2 | 4 | 680 | XP_011541538.1 | |
| XM_011543237.2 | 3 | 851 | XP_011541539.1 | |
| XM_017009166.1 | 4 | 867 | XP_016864655.1 | |
| XM_017009167.1 | 4 | 877 | XP_016864656.1 | |
| XM_017009168.1 | 5 | 596 | XP_016864657.1 | |
| XM_017009169.1 | 3 | 376 | XP_016864658.1 | |
| XM_017009170.1 | 3 | 801 | XP_016864659.1 | |
| AY358256.1 | 4 | 550 | AAQ88623.1 | |
| BC131563.1 | 4 | 482 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028027 | Chr.5:80398880 - 80545933 on Build GRCh38 | Gain |
|
| esv3605573 | Chr.5:80402118 - 80602933 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map