Assay Details
Target Gene Details
Entrez Gene ID: | 29985 |
Gene Name: | solute carrier family 39 member 3 |
Gene Aliases: |
ZIP-3, ZIP3 |
Location: |
Chr.19:2732524-2740076 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A3 | NM_144564.4 | NP_653165.2 | ||
| AF052125.1 | ||||
| AK302600.1 | ||||
| BC005869.2 | AAH05869.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509714 | Chr.19:2519629 - 2794403 on Build GRCh38 | Insertion |
|
| nsv828397 | Chr.19:2688987 - 2800506 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map