Assay Details
Target Gene Details
Entrez Gene ID: | 127247 |
Gene Name: | ankyrin repeat and SOCS box containing 17 |
Gene Aliases: |
Asb-17 |
Location: |
Chr.1:75918873-75937625 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB17 | NM_080868.2 | 1 | 67 | NP_543144.1 |
| NR_026546.1 | 1 | 67 | ||
| XM_006710345.3 | 1 | 43 | XP_006710408.1 | |
| XM_006710346.3 | 2 | 60 | XP_006710409.1 | |
| AF429968.1 | 1 | 45 | AAN85310.1 | |
| AK098606.1 | 1 | 67 | BAC05350.1 | |
| BC022517.1 | AAH22517.1 | |||
| BC036573.2 | 1 | 45 | AAH36573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004669 | Chr.1:75835513 - 75943766 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146637045] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map