Assay Details
Target Gene Details
Entrez Gene ID: | 645 |
Gene Name: | biliverdin reductase B |
Gene Aliases: |
BVRB, FLR, HEL-S-10, SDR43U1 |
Location: |
Chr.19:40447784-40465818 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLVRB | NM_000713.2 | 2 | 308 | NP_000704.1 |
| AK312137.1 | 2 | 235 | ||
| BC109371.1 | 2 | 225 | AAI09372.1 | |
| BG340491.1 | 2 | 308 | ||
| D26308.1 | 2 | 256 | BAA05370.1 | |
| D32143.1 | 2 | 287 | BAA06874.1 | |
| EU794593.1 | 2 | 308 |
More Information
Set Membership: |
|

Genomic Map