Assay Details
Target Gene Details
Entrez Gene ID: | 342574 |
Gene Name: | keratin 27 |
Gene Aliases: |
K25IRS3, KRT25C |
Location: |
Chr.17:40776808-40782534 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT27 | NM_181537.3 | NP_853515.2 | ||
| AJ564206.1 | CAD91906.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77854608] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map