Assay Details
Target Gene Details
Entrez Gene ID: | 7545 |
Gene Name: | Zic family member 1 |
Gene Aliases: |
CRS6, ZIC, ZNF201 |
Location: |
Chr.3:147409394-147416719 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZIC1 | NM_003412.3 | 3 | 4044 | NP_003403.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822285 | Chr.3:147381294 - 147428675 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map