Assay Details
Target Gene Details
Entrez Gene ID: | 6811 |
Gene Name: | syntaxin 5 |
Gene Aliases: |
SED5, STX5A |
Location: |
Chr.11:62806860-62832091 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STX5 | NM_001244666.1 | NP_001231595.1 | ||
| NM_003164.4 | NP_003155.2 | |||
| XM_011545223.2 | XP_011543525.1 | |||
| XM_011545224.2 | XP_011543526.1 | |||
| AK296675.1 | ||||
| AK313497.1 | ||||
| BC002645.1 | AAH02645.1 | |||
| BC012137.1 | AAH12137.2 | |||
| BT019646.1 | AAV38452.1 | |||
| BT019647.1 | AAV38453.1 | |||
| BX537426.1 | CAD97668.1 | |||
| U26648.1 | AAC71078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159903 | Chr.11:62818434 - 62885307 on Build GRCh38 | Duplication |
|
| nsv508636 | Chr.11:62779642 - 62876986 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map